Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001135044.1 | NP_001128516.1 |
NM_001308244.1 | NP_001295173.1 | |
NM_002752.4 | NP_002743.3 | |
NM_139068.2 | NP_620707.1 | |
NM_139069.2 | NP_620708.1 | |
NM_139070.2 | NP_620709.1 | |
XM_005265940.4 | XP_005265997.1 | |
XM_006714891.2 | XP_006714954.1 | |
XM_017009638.1 | XP_016865127.1 | |
XM_017009639.1 | XP_016865128.1 | |
XM_017009640.1 | XP_016865129.1 | |
XM_017009641.1 | XP_016865130.1 | |
XM_017009642.1 | XP_016865131.1 | |
XM_017009643.1 | XP_016865132.1 | |
GenBank mRNA | AB451302.1 | - |
AB451355.1 | - | |
AB451433.1 | - | |
AK289638.1 | - | |
BC032539.1 | - | |
BF056535.1 | - | |
BQ025589.1 | - | |
CR536580.1 | - | |
CX758487.1 | - | |
DB451562.1 | - | |
ENST00000393360.0 | - | |
EU927388.1 | - | |
HM036706.1 | - | |
HM036707.1 | - | |
HM036708.1 | - | |
HM036709.1 | - | |
L31951.1 | - | |
U09759.1 | - | |
U34821.1 | - | |
U35002.1 | - | |
U35003.1 | - |