Gene Transcripts

Gene Symbol
IKBKE
Entrez Gene ID
9641
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon
Gene Aliases
IKK-E, IKK-i, IKKE, IKKI
Chromosome Location
Chr.1: 206470243 - 206496890 on Build GRCh38
Uni Gene
Hs.321045
Interrogated Sequence Translated Protein
RefSeq NM_001193321.1 NP_001180250.1
NM_001193322.1 NP_001180251.1
NM_014002.3 NP_054721.1
XM_005273356.2 XP_005273413.1
XM_017002868.1 XP_016858357.1
GenBank mRNA AB016590.1 -
AF241789.1 -
AK093798.1 -
AK310737.1 -
AL706097.1 -
AY660028.1 -
BC105923.1 -
BC105924.1 -
BC107591.1 -
BC107812.1 -
CB243308.1 -
D63485.1 -
DA990116.1 -
DA999897.1 -
DB152295.1 -
ENST00000367120.0 -

View Full Product Details