Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001206389.1 | NP_001193318.1 |
NM_006119.4 | NP_006110.1 | |
NM_033163.3 | NP_149353.1 | |
NM_033164.3 | NP_149354.1 | |
NM_033165.3 | NP_149355.1 | |
GenBank mRNA | AB014615.1 | - |
BC069106.1 | - | |
BC128235.1 | - | |
BC128236.1 | - | |
CN369230.1 | - | |
ENST00000320185.0 | - | |
U36223.1 | - | |
U46211.1 | - | |
U46212.1 | - | |
U46213.1 | - | |
U56978.1 | - |