Gene Transcripts

Gene Symbol
AKT1
Entrez Gene ID
207
Gene Name
AKT serine/threonine kinase 1
Gene Aliases
AKT, CWS6, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
Chromosome Location
Chr.14: 104769349 - 104795743 on Build GRCh38
Uni Gene
Hs.525622
Interrogated Sequence Translated Protein
RefSeq NM_001014431.1 NP_001014431.1
NM_001014432.1 NP_001014432.1
NM_005163.2 NP_005154.2
XM_005267401.1 XP_005267458.1
XM_017021075.1 XP_016876564.1
XM_017021076.1 XP_016876565.1
XM_017021077.1 XP_016876566.1
XM_017021078.1 XP_016876567.1
GenBank mRNA AB451242.1 -
AB451367.1 -
AK094287.1 -
AK122894.1 -
AK131465.1 -
AK299310.1 -
AK314256.1 -
BC000479.2 -
BC001737.1 -
BC063408.1 -
BC084538.1 -
BX647722.1 -
BX648205.1 -
CN414251.1 -
ENST00000349310.0 -
KF836747.1 -
KF836748.1 -
KF836749.1 -
KF836750.1 -
KU177898.1 -
KU177899.1 -
M63167.1 -
X61037.1 -

View Full Product Details