Gene Transcripts

Gene Symbol
NGFR
Entrez Gene ID
4804
Gene Name
nerve growth factor receptor
Gene Aliases
CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTR
Chromosome Location
Chr.17: 49495293 - 49515020 on Build GRCh38
Uni Gene
Hs.415768
Interrogated Sequence Translated Protein
RefSeq NM_002507.3 NP_002498.1
GenBank mRNA AK303278.1 -
AK313654.1 -
BC050309.1 -
BC052797.1 -
BQ639556.1 -
CK299927.1 -
DA115173.1 -
ENST00000172229.0 -
M14764.1 -

View Full Product Details