Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001193321.1 | NP_001180250.1 |
NM_001193322.1 | NP_001180251.1 | |
NM_014002.3 | NP_054721.1 | |
XM_005273356.2 | XP_005273413.1 | |
XM_017002868.1 | XP_016858357.1 | |
GenBank mRNA | AB016590.1 | - |
AF241789.1 | - | |
AK093798.1 | - | |
AK310737.1 | - | |
AL706097.1 | - | |
AY660028.1 | - | |
BC105923.1 | - | |
BC105924.1 | - | |
BC107591.1 | - | |
BC107812.1 | - | |
CB243308.1 | - | |
D63485.1 | - | |
DA990116.1 | - | |
DA999897.1 | - | |
DB152295.1 | - | |
ENST00000367120.0 | - |