Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001198777.1 | NP_001185706.1 |
NM_001198778.1 | NP_001185707.1 | |
NM_001198779.1 | NP_001185708.1 | |
NM_001324375.1 | NP_001311304.1 | |
NM_001324376.1 | NP_001311305.1 | |
NM_003591.3 | NP_003582.2 | |
XM_011519743.1 | XP_011518045.1 | |
XM_011519744.1 | XP_011518046.1 | |
XM_011519745.1 | XP_011518047.1 | |
XM_011519747.1 | XP_011518049.1 | |
GenBank mRNA | AF126404.1 | - |
AI089201.1 | - | |
AK095217.1 | - | |
AK294080.1 | - | |
AK300491.1 | - | |
AK310225.1 | - | |
AL832733.1 | - | |
BC009591.1 | - | |
BC095405.1 | - | |
BC110901.1 | - | |
DC401811.1 | - | |
DC401936.1 | - | |
ENST00000374751.0 | - | |
U58088.1 | - | |
U83410.1 | - |