Gene Transcripts

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
Chromosome Location
Chr.7: 81699006 - 81770438 on Build GRCh38
Uni Gene
Hs.396530
Interrogated Sequence Translated Protein
RefSeq NM_000601.5 NP_000592.3
NM_001010931.2 NP_001010931.1
NM_001010932.2 NP_001010932.1
NM_001010933.2 NP_001010933.1
NM_001010934.2 NP_001010934.1
XM_006715956.2 XP_006716019.1
XM_011516115.2 XP_011514417.1
XM_017012097.1 XP_016867586.1
XM_017012098.1 XP_016867587.1
GenBank mRNA AB208900.1 -
AK291674.1 -
AK313694.1 -
AV701693.1 -
BC022308.1 -
BC063485.1 -
BC105797.1 -
BC130284.1 -
BC130286.1 -
BM153501.1 -
BM971901.1 -
BU533235.1 -
BX495370.1 -
ENST00000222390.0 -
FJ830861.1 -
FJ830862.1 -
L02931.1 -
M29145.1 -
M55379.1 -
M60718.1 -
M73239.1 -
M73240.1 -
M77227.1 -
U46010.1 -
X16323.1 -
X57574.1 -

View Full Product Details