Gene Transcripts

Gene Symbol
EGFR
Entrez Gene ID
1956
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
Chromosome Location
Chr.7: 55019032 - 55207338 on Build GRCh38
Uni Gene
Hs.488293
Interrogated Sequence Translated Protein
RefSeq NM_005228.3 NP_005219.2
NM_201282.1 NP_958439.1
NM_201283.1 NP_958440.1
NM_201284.1 NP_958441.1
GenBank mRNA AB209442.1 -
AF125253.1 -
AF277897.1 -
AI217671.1 -
AK127817.1 -
AK225422.1 -
AK290352.1 -
AK294750.1 -
AL598260.1 -
AU137334.1 -
AW163038.1 -
AW295229.1 -
AY550966.1 -
AY698023.1 -
AY698024.1 -
BC070081.1 -
BC094761.1 -
BC118665.1 -
BC128419.1 -
CB160831.1 -
DQ088980.1 -
EF210210.1 -
EF210211.1 -
ENST00000275493.0 -
GU228573.1 -
GU228584.1 -
GU255993.1 -
HM437234.1 -
HM437235.1 -
HQ730875.1 -
HQ912715.1 -
HQ999997.1 -
K03193.1 -
U48722.1 -
U51732.1 -
U95089.1 -
X00588.1 -
X00663.1 -

View Full Product Details