Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_019111.4 | NP_061984.2 |
GenBank mRNA | AI492718.1 | - |
AK297032.1 | - | |
AK301134.1 | - | |
AK301190.1 | - | |
AK301354.1 | - | |
AK304429.1 | - | |
AK310161.1 | - | |
AK313123.1 | - | |
BC032350.1 | - | |
BC071659.1 | - | |
BG548634.1 | - | |
BM849755.1 | - | |
CR457013.1 | - | |
ENST00000395388.0 | - | |
J00193.1 | - | |
J00194.1 | - | |
J00195.1 | - | |
J00197.1 | - | |
K01171.1 | - | |
M35979.1 | - | |
M60333.1 | - | |
M60334.1 | - | |
V00523.1 | - | |
V00528.1 | - |