Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001278599.1 | NP_001265528.1 |
NM_001699.5 | NP_001690.2 | |
NM_021913.4 | NP_068713.2 | |
GenBank mRNA | AA021223.1 | - |
AI300772.1 | - | |
AI309979.1 | - | |
AK309139.1 | - | |
AK310881.1 | - | |
AL556721.3 | - | |
BC032229.1 | - | |
CA437238.1 | - | |
DA478638.1 | - | |
ENST00000359092.0 | - | |
HY045208.1 | - | |
M76125.1 | - | |
S65125.1 | - | |
X57019.1 | - | |
X66029.1 | - |