Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001251825.1 | NP_001238754.1 |
NM_003109.1 | NP_003100.1 | |
NM_138473.2 | NP_612482.2 | |
XM_011538696.2 | XP_011536998.1 | |
GenBank mRNA | AB209678.1 | - |
AF252284.1 | - | |
AF255682.1 | - | |
AI282745.1 | - | |
AI561005.1 | - | |
AJ272134.1 | - | |
AK311995.1 | - | |
AL442093.1 | - | |
AW028976.1 | - | |
BC012008.1 | - | |
BC043224.1 | - | |
BC062539.1 | - | |
BC078158.1 | - | |
BF434304.1 | - | |
BF436290.1 | - | |
BF671559.1 | - | |
BM463205.1 | - | |
BQ072120.1 | - | |
BQ431320.1 | - | |
BQ774060.1 | - | |
CD104746.1 | - | |
FJ949572.1 | - | |
FN908228.1 | - | |
J03133.1 | - |