Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001099691.2 | NP_001093161.1 |
NM_001308158.1 | NP_001295087.1 | |
NM_001308159.1 | NP_001295088.1 | |
NM_003236.3 | NP_003227.1 | |
GenBank mRNA | AF149096.1 | - |
AF149097.1 | - | |
AF149098.1 | - | |
AK290151.1 | - | |
AK309066.1 | - | |
AK309220.1 | - | |
AK312899.1 | - | |
AY548763.1 | - | |
BC005308.1 | - | |
BI760767.1 | - | |
BM453873.1 | - | |
BT006833.1 | - | |
CD369689.1 | - | |
CN274505.1 | - | |
DA216480.1 | - | |
DA229819.1 | - | |
DA373586.1 | - | |
DA462271.1 | - | |
ENST00000295400.0 | - | |
K03222.1 | - | |
M31172.1 | - | |
X70340.1 | - |