Gene Transcripts

Gene Symbol
Zap70
Entrez Gene ID
22637
Gene Name
zeta-chain (TCR) associated protein kinase
Gene Aliases
Srk, ZAP-70, mrtle, mur
Chromosome Location
Chr.1: 36761798 - 36782821 on Build GRCm38
Uni Gene
Mm.8038
Interrogated Sequence Translated Protein
RefSeq NM_001289612.1 NP_001276541.1
NM_001289765.1 NP_001276694.1
NM_001289766.1 NP_001276695.1
NM_009539.3 NP_033565.2
GenBank mRNA AB083210.1 -
AB084383.1 -
AK039883.1 -
AK089056.1 -
BC029727.1 -
BY589032.1 -
CJ143180.1 -
U04379.1 -
U77667.1 -

View Full Product Details