Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001289612.1 | NP_001276541.1 |
NM_001289765.1 | NP_001276694.1 | |
NM_001289766.1 | NP_001276695.1 | |
NM_009539.3 | NP_033565.2 | |
GenBank mRNA | AB083210.1 | - |
AB084383.1 | - | |
AK039883.1 | - | |
AK089056.1 | - | |
BC029727.1 | - | |
BY589032.1 | - | |
CJ143180.1 | - | |
U04379.1 | - | |
U77667.1 | - |