Gene Transcripts

Gene Symbol
Mertk
Entrez Gene ID
17289
Gene Name
c-mer proto-oncogene tyrosine kinase
Gene Aliases
Eyk, Mer, Nyk, nmf12
Chromosome Location
Chr.2: 128698997 - 128803162 on Build GRCm38
Uni Gene
Mm.239655
Interrogated Sequence Translated Protein
RefSeq NM_008587.1 NP_032613.1
GenBank mRNA AK029009.1 -
AK030158.1 -
AK079252.1 -
L11625.1 -
U21301.1 -

View Full Product Details