Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001163671.1 | NP_001157143.1 |
NM_001163672.1 | NP_001157144.1 | |
NM_016961.3 | NP_058657.1 | |
NM_207692.2 | NP_997575.2 | |
GenBank mRNA | AB005664.1 | - |
AF052466.1 | - | |
AF052467.1 | - | |
AF052468.1 | - | |
AF052469.1 | - | |
AK031959.1 | - | |
AK031974.1 | - | |
AK043169.1 | - | |
AK090057.1 | - | |
AK145083.1 | - | |
BB362351.1 | - | |
BC024514.1 | - | |
BC028341.1 | - | |
CX235052.1 | - |