Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_029402.3 | NP_083678.2 |
GenBank mRNA | AK004875.2 | - |
AK016520.1 | - | |
AK028589.1 | - | |
AK144693.1 | - | |
AK160597.1 | - | |
AK162910.1 | - | |
AK204549.1 | - | |
AK220287.1 | - | |
BC025902.1 | - | |
BC026779.1 | - | |
BC027428.1 | - |