Gene Transcripts

Gene Symbol
Ikbke
Entrez Gene ID
56489
Gene Name
inhibitor of kappaB kinase epsilon
Gene Aliases
AW558201, IKK-i, IKKepsilon, Ikki
Chromosome Location
Chr.1: 131254600 - 131279625 on Build GRCm38
Uni Gene
Mm.386783
Interrogated Sequence Translated Protein
RefSeq NM_019777.3 NP_062751.2
GenBank mRNA AB016589.1 -
AK088580.1 -
AK149911.1 -
AK153046.1 -
AK180824.1 -
AK193494.1 -
AK198423.1 -
AK209824.1 -
AK213678.1 -
AK218764.1 -
AK220324.1 -
BC037446.1 -
CT010206.1 -
CT010313.1 -

View Full Product Details