Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_019777.3 | NP_062751.2 |
GenBank mRNA | AB016589.1 | - |
AK088580.1 | - | |
AK149911.1 | - | |
AK153046.1 | - | |
AK180824.1 | - | |
AK193494.1 | - | |
AK198423.1 | - | |
AK209824.1 | - | |
AK213678.1 | - | |
AK218764.1 | - | |
AK220324.1 | - | |
BC037446.1 | - | |
CT010206.1 | - | |
CT010313.1 | - |