Gene Transcripts

Gene Symbol
Il12b
Entrez Gene ID
16160
Gene Name
interleukin 12b
Gene Aliases
Il-12b, Il-12p40, Il12p40, p40
Chromosome Location
Chr.11: 44400063 - 44414677 on Build GRCm38
Uni Gene
Mm.239707
Interrogated Sequence Translated Protein
GenBank mRNA AF128214.1 -
AF128215.1 -
AK162981.1 -
BC103608.1 -
BC103609.1 -
BC103610.1 -
BC103614.1 -
BY154173.1 -
M86671.1 -

View Full Product Details