Gene Transcripts

Gene Symbol
Ctla4
Entrez Gene ID
12477
Gene Name
cytotoxic T-lymphocyte-associated protein 4
Gene Aliases
Cd152, Ctla-4, Ly-56
Chromosome Location
Chr.1: 60909025 - 60915832 on Build GRCm38
Uni Gene
Mm.390
Interrogated Sequence Translated Protein
RefSeq NM_001281976.1 NP_001268905.1
NM_009843.4 NP_033973.2
GenBank mRNA AB097214.1 -
AI323635.1 -
AK138891.1 -
BC042741.1 -
BC052683.1 -
U90270.1 -
X05719.1 -

View Full Product Details