Gene Transcripts

Gene Symbol
Gdf15
Entrez Gene ID
23886
Gene Name
growth differentiation factor 15
Gene Aliases
MIC-1, NAG-1, SBF
Chromosome Location
Chr.8: 70629393 - 70632469 on Build GRCm38
Uni Gene
Mm.31325
Interrogated Sequence Translated Protein
RefSeq NM_011819.2 NP_035949.2
GenBank mRNA AF159571.1 -
AK144199.1 -
AK159242.1 -
AK160927.1 -
BC067248.1 -

View Full Product Details