Gene Transcripts

Gene Symbol
Mapk9
Entrez Gene ID
26420
Gene Name
mitogen-activated protein kinase 9
Gene Aliases
AI851083, JNK2, Prkm9, p54aSAPK
Chromosome Location
Chr.11: 49846751 - 49886421 on Build GRCm38
Uni Gene
Mm.68933
Interrogated Sequence Translated Protein
RefSeq NM_001163671.1 NP_001157143.1
NM_001163672.1 NP_001157144.1
NM_016961.3 NP_058657.1
NM_207692.2 NP_997575.2
GenBank mRNA AB005664.1 -
AF052466.1 -
AF052467.1 -
AF052468.1 -
AF052469.1 -
AK031959.1 -
AK031974.1 -
AK043169.1 -
AK090057.1 -
AK145083.1 -
BB362351.1 -
BC024514.1 -
BC028341.1 -
CX235052.1 -

View Full Product Details