Gene Transcripts

Gene Symbol
Bad
Entrez Gene ID
12015
Gene Name
BCL2-associated agonist of cell death
Gene Aliases
AI325008, Bbc2
Chromosome Location
Chr.19: 6941861 - 6951905 on Build GRCm38
Uni Gene
Mm.4387
Interrogated Sequence Translated Protein
RefSeq NM_001285453.1 NP_001272382.1
NM_007522.3 NP_031548.1
GenBank mRNA AK029400.1 -
AK131666.1 -
AK151792.1 -
AK169002.1 -
BC006762.1 -
L37296.1 -

View Full Product Details