Gene Transcripts

Gene Symbol
Ikbkg
Entrez Gene ID
16151
Gene Name
inhibitor of kappaB kinase gamma
Gene Aliases
1110037D23Rik, AI848108, AI851264, AW124339, IKK[g], NEMO
Chromosome Location
Chr.X: 74393291 - 74456792 on Build GRCm38
Uni Gene
Mm.12967
Interrogated Sequence Translated Protein
RefSeq NM_001136067.2 NP_001129539.1
NM_001161421.1 NP_001154893.1
NM_001161422.1 NP_001154894.1
NM_001161423.1 NP_001154895.1
NM_001161424.1 NP_001154896.1
NM_010547.2 NP_034677.2
NM_178590.4 NP_848705.1
GenBank mRNA AF069542.1 -
AF513109.1 -
AK037020.1 -
AK042138.1 -
AK047400.1 -
AK048446.1 -
AK148163.1 -
AK154095.1 -
AK188269.1 -
AY112937.1 -
BC021431.1 -
BC026905.1 -
BY768708.1 -
CJ099503.1 -
CN701499.1 -
CN717929.1 -
CV556906.1 -

View Full Product Details