Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001136067.2 | NP_001129539.1 |
NM_001161421.1 | NP_001154893.1 | |
NM_001161422.1 | NP_001154894.1 | |
NM_001161423.1 | NP_001154895.1 | |
NM_001161424.1 | NP_001154896.1 | |
NM_010547.2 | NP_034677.2 | |
NM_178590.4 | NP_848705.1 | |
GenBank mRNA | AF069542.1 | - |
AF513109.1 | - | |
AK037020.1 | - | |
AK042138.1 | - | |
AK047400.1 | - | |
AK048446.1 | - | |
AK148163.1 | - | |
AK154095.1 | - | |
AK188269.1 | - | |
AY112937.1 | - | |
BC021431.1 | - | |
BC026905.1 | - | |
BY768708.1 | - | |
CJ099503.1 | - | |
CN701499.1 | - | |
CN717929.1 | - | |
CV556906.1 | - |