Gene Transcripts

Gene Symbol
Map3k3
Entrez Gene ID
26406
Gene Name
mitogen-activated protein kinase kinase kinase 3
Gene Aliases
AW548911, MAPKKK3, Mekk3, mKIAA4031
Chromosome Location
Chr.11: 106084902 - 106156656 on Build GRCm38
Uni Gene
Mm.27041
Interrogated Sequence Translated Protein
RefSeq NM_011947.3 NP_036077.1
GenBank mRNA AK137127.1 -
AK142278.1 -
AK220450.1 -
BC023781.1 -
U43187.1 -

View Full Product Details