Gene Transcripts
- Gene Symbol
- YBX3P1
- Entrez Gene ID
- 440359
- Gene Name
- Y-box binding protein 3 pseudogene 1
- Gene Aliases
- CSDAP1
- Chromosome Location
- Chr. 16: 31567767 - 31569524 on Build GRCh38
- UniGene ID
- Hs.650902
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027011.1 |
|
1 |
375 |
|
GenBank mRNA |
BI826429.1 |
- |
1 |
83 |
|
DB451883.1 |
- |
1 |
346 |
|
View Full Product Details