Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001322177.1 | NP_001309106.1 | 2 | 4964 | |
NM_177966.6 | NP_808881.3 | 3 | 4791 | ||
GenBank mRNA | U79276.1 | - | 1 | 926 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | EB386946.1 | - | 1 | 635 | Chr. 3 - 57562198 - 57563037 [+] |