Gene Transcripts
- Gene Symbol
- MCF2L-AS1
- Entrez Gene ID
- 100289410
- Gene Name
- MCF2L antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 13: 112967484 - 112968638 on Build GRCh38
- UniGene ID
- Hs.667436
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034002.1 |
|
1 |
854 |
|
GenBank mRNA |
BC064982.1 |
- |
|
975 |
|
BC080517.1 |
- |
|
975 |
|
View Full Product Details