Gene Transcripts

Gene Symbol
MLEC
Entrez Gene ID
9761
Gene Name
malectin
Gene Aliases
KIAA0152
Chromosome Location
Chr. 12: 120687124 - 120701864 on Build GRCh38
UniGene ID
Hs.744910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011539031.1 XP_011537333.1 1 2122
XM_011539032.1 XP_011537334.1 1 2122
GenBank mRNA AK124275.1 - 1 1700

View Full Product Details