Gene Transcripts

Gene Symbol
ZNF101
Entrez Gene ID
94039
Gene Name
zinc finger protein 101
Gene Aliases
HZF12
Chromosome Location
Chr. 19: 19668153 - 19683506 on Build GRCh38
UniGene ID
Hs.631642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300949.1 NP_001287878.1 4 2819
NM_033204.3 NP_149981.2 4 2947
GenBank mRNA AK123904.1 - 1 534

View Full Product Details