Gene Transcripts

Gene Symbol
NAALADL2
Entrez Gene ID
254827
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 2
Gene Aliases
Chromosome Location
Chr. 3: 174439669 - 175809510 on Build GRCh38
UniGene ID
Hs.565848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006713560.2 XP_006713623.1 16 7868
XM_011512612.2 XP_011510914.1 14 11122
XM_011512613.1 XP_011510915.1 14 7930
XM_011512615.2 XP_011510917.1 13 10979
XM_011512616.2 XP_011510918.1 13 8660
XM_017006070.1 XP_016861559.1 18 8310
XM_017006071.1 XP_016861560.1 19 8380
XM_017006072.1 XP_016861561.1 20 8514
XM_017006073.1 XP_016861562.1 18 8113
XM_017006074.1 XP_016861563.1 20 10532
XM_017006075.1 XP_016861564.1 17 9257
XM_017006076.1 XP_016861565.1 17 8050
XM_017006077.1 XP_016861566.1 19 9461
XM_017006078.1 XP_016861567.1 17 9278
XM_017006079.1 XP_016861568.1 15 8754
XM_017006080.1 XP_016861569.1 15 9125
XM_017006081.1 XP_016861570.1 13 10975
XM_017006082.1 XP_016861571.1 15 8973
XM_017006083.1 XP_016861572.1 14 7414
GenBank mRNA AK021754.1 - 1 727

View Full Product Details