Gene Transcripts
- Gene Symbol
- E230015B07Rik
- Entrez Gene ID
- 320001
- Gene Name
- RIKEN cDNA E230015B07 gene
- Gene Aliases
- Chromosome Location
- Chr. 6: 87672370 - 87679920 on Build GRCm38
- UniGene ID
- Mm.137379
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042791.1 |
- |
1, 2 |
198 |
|
AK054052.1 |
- |
|
148 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB639992.1 |
- |
1, 2 |
198 |
|
Chr. 6 - 87672382 - 87679135 [+] |
GenBankmRNA |
NM_177057.1 |
- |
|
148 |
|
Chr. - - [-] |
View Full Product Details