Gene Transcripts
- Gene Symbol
- Gm13293
- Entrez Gene ID
- 100503147
- Gene Name
- predicted gene 13293
- Gene Aliases
- OTTMUSG00000011311
- Chromosome Location
- Chr. 2: 11339489 - 11344106 on Build GRCm38
- UniGene ID
- Mm.91961
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040369.1 |
|
2, 3 |
499 |
|
GenBank mRNA |
AK140161.1 |
- |
2, 3 |
499 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY727245.1 |
- |
2, 3 |
499 |
|
Chr. 2 - 11339489 - 11343170 [+] |
View Full Product Details