Gene Transcripts

Gene Symbol
2010107E04Rik
Entrez Gene ID
70257
Gene Name
RIKEN cDNA 2010107E04 gene
Gene Aliases
9430003J03Rik, AU043134, AV124504, Mp68
Chromosome Location
Chr. 12: 111961375 - 111966977 on Build GRCm38
UniGene ID
Mm.318
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK020400.1 - 1 216


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CA751014.1 - 1 588 Chr. 12 - 111963401 - 111964176 [-]

View Full Product Details