Gene Transcripts

Gene Symbol
Mib1
Entrez Gene ID
225164
Gene Name
mindbomb homolog 1 (Drosophila)
Gene Aliases
DIP-1, E430019M12Rik, Mib, mKIAA1323
Chromosome Location
Chr. 18: 10725625 - 10818578 on Build GRCm38
UniGene ID
Mm.21500
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK080083.1 - 1 1091
AK080847.1 - 15 3808
AK088539.1 - 1 3480
AK153523.1 - 1 178
AK161848.1 - 7 2553
BC083072.1 - 11 6718

View Full Product Details