Gene Transcripts

Gene Symbol
Myl6b
Entrez Gene ID
216459
Gene Name
myosin, light polypeptide 6B
Gene Aliases
5730437E04Rik, BC037527, MLC1SA
Chromosome Location
Chr. 10: 128494157 - 128498685 on Build GRCm38
UniGene ID
Mm.291688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017625.1 - 1 933


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AI640003.1 - 1 164 Chr. 10 - 128495082 - 128495586 [+]
GenBankmRNA BY528579.1 - 1 54 Chr. 10 - 128495192 - 128495556 [+]
GenBankmRNA BY383433.1 - 1 97 Chr. 10 - 128495149 - 128495560 [+]
GenBankmRNA DV047970.1 - 1 501 Chr. 10 - 128494763 - 128495555 [+]
GenBankmRNA AA162263.1 - 1 163 Chr. 10 - 128495083 - 128495538 [+]

View Full Product Details