Gene Transcripts
- Gene Symbol
- Lipt1
- Entrez Gene ID
- 623661
- Gene Name
- lipoyltransferase 1
- Gene Aliases
- EG623661
- Chromosome Location
- Chr. 1: 37868507 - 37876298 on Build GRCm38
- UniGene ID
- Mm.138000
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK039030.1 |
- |
|
973 |
|
View Full Product Details