Gene Transcripts
- Gene Symbol
- 2310069B03Rik
- Entrez Gene ID
- 69652
- Gene Name
- RIKEN cDNA 2310069B03 gene
- Gene Aliases
- Chromosome Location
- Chr. 6: 82877866 - 82881853 on Build GRCm38
- UniGene ID
- Mm.96357
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040520.1 |
|
1, 2 |
88 |
|
GenBank mRNA |
AK078808.1 |
- |
1, 2 |
89 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB624608.1 |
- |
1, 2 |
85 |
|
Chr. 6 - 82879064 - 82881850 [-] |
View Full Product Details