Gene Transcripts
- Gene Symbol
- Xrcc4
- Entrez Gene ID
- 108138
- Gene Name
- X-ray repair complementing defective repair in Chinese hamster cells 4
- Gene Aliases
- 2310057B22Rik, AW413319, AW545101
- Chromosome Location
- Chr. 13: 89848904 - 90089608 on Build GRCm38
- UniGene ID
- Mm.37531
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038105.1 |
- |
1 |
44 |
|
View Full Product Details