Gene Transcripts
- Gene Symbol
- LINC01487
- Entrez Gene ID
- 101928190
- Gene Name
- long intergenic non-protein coding RNA 1487
- Gene Aliases
- Chromosome Location
- Chr. 3: 155240945 - 155243124 on Build GRCh38
- UniGene ID
- Hs.121410
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_125399.1 |
|
3 |
295 |
|
GenBank mRNA |
BU570602.1 |
- |
3 |
295 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000462531.1 |
- |
2 |
794 |
|
Chr. 3 - 155240945 - 155243124 [+] |
View Full Product Details