Gene Transcripts

Gene Symbol
Mark1
Entrez Gene ID
226778
Gene Name
MAP/microtubule affinity regulating kinase 1
Gene Aliases
AW491150, B930025N23Rik, Emk3, Par-1c, mKIAA1477, mPar-1c
Chromosome Location
Chr. 1: 184896424 - 184999572 on Build GRCm38
UniGene ID
Mm.7445
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145515.2 NP_663490.2 18 2404
GenBank mRNA AB185282.1 - 18 2330
AF453686.1 - 18 2334
AK173181.1 - 17 2257
BC112400.1 - 18 2333 40059725

View Full Product Details