Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145569.4 | NP_663544.1 | 5 | 626 | |
GenBank mRNA | AB070266.1 | - | 5 | 590 | |
AK034757.1 | - | 5 | 627 | ||
AK052764.1 | - | 5 | 625 | ||
AK079420.1 | - | 5 | 632 | ||
AK133960.1 | - | 5 | 625 | ||
AK145407.1 | - | 5 | 625 | ||
AK150040.1 | - | 5 | 625 | ||
AK151281.1 | - | 5 | 625 | ||
AK160869.1 | - | 5 | 625 | ||
AK161852.1 | - | 5 | 625 | ||
AK164137.1 | - | 5 | 625 | ||
AK166671.1 | - | 5 | 625 | ||
AK168155.1 | - | 5 | 625 | ||
AK169705.1 | - | 5 | 625 | ||
BC003451.1 | - | 5 | 566 | 2655444 | |
BC058360.1 | - | 5 | 607 | 6821781 |