Gene Transcripts

Gene Symbol
Lrrc3
Entrez Gene ID
237387
Gene Name
leucine rich repeat containing 3
Gene Aliases
1300011L04Rik, C21orf102
Chromosome Location
Chr. 10: 77897575 - 77902536 on Build GRCm38
UniGene ID
Mm.133301
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145152.4 NP_660134.1 2 981
GenBank mRNA AK043535.1 - 2 956
AY061858.1 - 2 954
BC115652.2 - 1 789 40103158

View Full Product Details