Gene Transcripts

Gene Symbol
Leap2
Entrez Gene ID
259301
Gene Name
liver-expressed antimicrobial peptide 2
Gene Aliases
leap-2
Chromosome Location
Chr. 11: 53422181 - 53423136 on Build GRCm38
UniGene ID
Mm.24283
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153069.3 NP_694709.1 2 118
GenBank mRNA AJ409055.1 - 2 118
BC089593.1 - 2 111 6749700
W82161.1 - 2 152

View Full Product Details