Gene Transcripts

Gene Symbol
Myo9a
Entrez Gene ID
270163
Gene Name
myosin IXa
Gene Aliases
4732465J09Rik, C130068I12Rik, C230003M11
Chromosome Location
Chr. 9: 59750856 - 59928866 on Build GRCm38
UniGene ID
Mm.249545
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173018.2 NP_766606.2 37 6833
GenBank mRNA AK028873.1 - 8 836
AK029836.1 - 7 700
BC068298.1 - 11 1268

View Full Product Details