Gene Transcripts

Gene Symbol
Lipt1
Entrez Gene ID
623661
Gene Name
lipoyltransferase 1
Gene Aliases
EG623661
Chromosome Location
Chr. 1: 37868507 - 37876298 on Build GRCm38
UniGene ID
Mm.138000
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037918.3 NP_001033007.2 2 977
GenBank mRNA AK048005.1 - 978
BC019519.1 - 2 966 4038179

View Full Product Details