Gene Transcripts

Gene Symbol
Epo
Entrez Gene ID
24335
Gene Name
erythropoietin
Gene Aliases
Chromosome Location
Chr. 12: 22274828 - 22278268 on Build Rnor_6.0
UniGene ID
Rn.11365
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017001.1 NP_058697.1 4 381
XM_006249101.2 XP_006249163.1 3 523
GenBank mRNA D10763.1 - 4 381
L10608.1 - 3 350

View Full Product Details